Anti-NFE2L2 Mouse Monoclonal Antibody [clone: 383727]

Supplier: R & D Systems
MAB3925
220020-674EA 655.11 USD
220020-674
Anti-NFE2L2 Mouse Monoclonal Antibody [clone: 383727]
Antibodies
Type: Primary
Antigen Symbol: NFE2L2
Antigen Name: nuclear factor, erythroid 2-like 2
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 383727
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR