Anti-MAFG Mouse Monoclonal Antibody [clone: 370412]

Supplier: R & D Systems
MAB3924
220020-673EA 577.02 USD
220020-673
Anti-MAFG Mouse Monoclonal Antibody [clone: 370412]
Antibodies
Type: Primary
Antigen: MAFG
Clonality: Monoclonal
Clone: 370412
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR