Anti-RALB Rat Monoclonal Antibody [clone: 399417]

Supplier: R & D Systems
MAB3920
220020-672EA 577.02 USD
220020-672
Anti-RALB Rat Monoclonal Antibody [clone: 399417]
Antibodies
Type: Primary
Antigen: RALB
Clonality: Monoclonal
Clone: 399417
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR