Anti-RPS6KA6 Rat Monoclonal Antibody [clone: 374521]

Supplier: R & D Systems
MAB3916
220020-670EA 566.89 USD
220020-670
Anti-RPS6KA6 Rat Monoclonal Antibody [clone: 374521]
Antibodies
Type: Primary
Antigen: RPS6KA6
Clonality: Monoclonal
Clone: 374521
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR