Anti-HCK Rat Monoclonal Antibody [clone: 394903]

Supplier: R & D Systems
MAB3915
220020-669EA 594.37 USD
220020-669
Anti-HCK Rat Monoclonal Antibody [clone: 394903]
Antibodies
Type: Primary
Antigen Symbol: HCK
Antigen Name: HCK proto-oncogene, Src family tyrosine kinase
Antigen Synonyms: JTK9, p59Hck, p61Hck
Clonality: Monoclonal
Clone: 394903
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR