Anti-MAPK4 Rat Monoclonal Antibody [clone: 355810]

Supplier: R & D Systems
MAB3914
220020-668EA 594.37 USD
220020-668
Anti-MAPK4 Rat Monoclonal Antibody [clone: 355810]
Antibodies
Type: Primary
Antigen: MAPK4
Clonality: Monoclonal
Clone: 355810
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR