Anti-AICDA Rat Monoclonal Antibody [clone: 359218]

Supplier: R & D Systems
MAB39102
102885-660EA 577.02 USD
102885-660
Anti-AICDA Rat Monoclonal Antibody [clone: 359218]
Antibodies
Type: Primary
Antigen: AICDA
Clonality: Monoclonal
Clone: 359218
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR