Anti-DPP4 Mouse Monoclonal Antibody [clone: 354020]

Supplier: R & D Systems
MAB3908
220020-666EA 594.37 USD
220020-666
Anti-DPP4 Mouse Monoclonal Antibody [clone: 354020]
Antibodies
Type: Primary
Antigen: DPP4
Clonality: Monoclonal
Clone: 354020
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR