Anti-CD160 Rat Monoclonal Antibody [clone: 342705]

Supplier: R & D Systems
MAB38991
102885-658EA 577.02 USD
102885-658
Anti-CD160 Rat Monoclonal Antibody [clone: 342705]
Antibodies
Type: Primary
Antigen: CD160
Clonality: Monoclonal
Clone: 342705
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR