Anti-CALR Mouse Monoclonal Antibody [clone: 681233]

Supplier: R & D Systems
MAB38981
102885-656EA 616.06 USD
102885-656
Anti-CALR Mouse Monoclonal Antibody [clone: 681233]
Antibodies
Type: Primary
Antigen: CALR
Clonality: Monoclonal
Clone: 681233
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR