Anti-SLC27A5 Mouse Monoclonal Antibody [clone: 390917]

Supplier: R & D Systems
MAB3897
220020-663EA 577.02 USD
220020-663
Anti-SLC27A5 Mouse Monoclonal Antibody [clone: 390917]
Antibodies
Type: Primary
Antigen: SLC27A5
Clonality: Monoclonal
Clone: 390917
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR