Anti-OPRK1 Mouse Monoclonal Antibody [clone: 387310]

Supplier: R & D Systems
MAB38951
220020-661EA 577.02 USD
220020-661
Anti-OPRK1 Mouse Monoclonal Antibody [clone: 387310]
Antibodies
Type: Primary
Antigen: OPRK1
Clonality: Monoclonal
Clone: 387310
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR