Anti-PCSK9 Rat Monoclonal Antibody [clone: 499911]

Supplier: R & D Systems
MAB38882
220020-658EA 616.06 USD
220020-658
Anti-PCSK9 Rat Monoclonal Antibody [clone: 499911]
Antibodies
Type: Primary
Antigen: PCSK9
Clonality: Monoclonal
Clone: 499911
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR