Anti-PCSK9 Mouse Monoclonal Antibody [clone: 410420]

Supplier: R & D Systems
MAB38881
220020-657EA 594.37 USD
220020-657
Anti-PCSK9 Mouse Monoclonal Antibody [clone: 410420]
Antibodies
Type: Primary
Antigen: PCSK9
Clonality: Monoclonal
Clone: 410420
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR