Anti-PCSK9 Rat Monoclonal Antibody [clone: 407119]

Supplier: R & D Systems
MAB3888
220020-656EA 633.42 USD
220020-656
Anti-PCSK9 Rat Monoclonal Antibody [clone: 407119]
Antibodies
Type: Primary
Antigen: PCSK9
Clonality: Monoclonal
Clone: 407119
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR