Anti-PLXNC1 Mouse Monoclonal Antibody [clone: 544232]

Supplier: R & D Systems
MAB3887
102885-654EA 642.09 USD
102885-654
Anti-PLXNC1 Mouse Monoclonal Antibody [clone: 544232]
Antibodies
Type: Primary
Antigen: PLXNC1
Clonality: Monoclonal
Clone: 544232
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR