Anti-VTN Rat Monoclonal Antibody [clone: 347317]

Supplier: R & D Systems
MAB38751
220020-653EA 600.15 USD
220020-653
Anti-VTN Rat Monoclonal Antibody [clone: 347317]
Antibodies
Type: Primary
Antigen: VTN
Clonality: Monoclonal
Clone: 347317
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR