Anti-VTN Rat Monoclonal Antibody [clone: 347312]

Supplier: R & D Systems
MAB3875
220020-652EA 577.02 USD
220020-652
Anti-VTN Rat Monoclonal Antibody [clone: 347312]
Antibodies
Type: Primary
Antigen: VTN
Clonality: Monoclonal
Clone: 347312
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR