Anti-NRTN Mouse Monoclonal Antibody [clone: 126424]

Supplier: R & D Systems
MAB3871
220020-651EA 772.25 USD
220020-651
Anti-NRTN Mouse Monoclonal Antibody [clone: 126424]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: NRTN
Clonality: Monoclonal
Clone: 126424
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR