Anti-NRP1 Mouse Monoclonal Antibody [clone: 446921]

Supplier: R & D Systems
MAB3870
220020-649EA 655.11 USD
220020-649
Anti-NRP1 Mouse Monoclonal Antibody [clone: 446921]
Antibodies
Type: Primary
Antigen: NRP1
Clonality: Monoclonal
Clone: 446921
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR