Anti-NRTN Mouse Monoclonal Antibody [clone: 126422]

Supplier: R & D Systems
MAB387
220020-648EA 763.57 USD
220020-648
Anti-NRTN Mouse Monoclonal Antibody [clone: 126422]
Antibodies
Blockade of Receptor-ligand Interaction

Type: Primary
Antigen: NRTN
Clonality: Monoclonal
Clone: 126422
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR