Anti-FGF22 Mouse Monoclonal Antibody [clone: 435008]

Supplier: R & D Systems
MAB3867
220020-647EA 763.57 USD
220020-647
Anti-FGF22 Mouse Monoclonal Antibody [clone: 435008]
Antibodies
Type: Primary
Antigen Symbol: FGF22
Antigen Name: fibroblast growth factor 22
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 435008
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR