Anti-B2M Mouse Monoclonal Antibody [clone: 747502]

Supplier: R & D Systems
MAB3864
102885-650EA 561.11 USD
102885-650
Anti-B2M Mouse Monoclonal Antibody [clone: 747502]
Antibodies
Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: 747502
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR