Anti-FYB Mouse Monoclonal Antibody [clone: 460107]

Supplier: R & D Systems
MAB38571
220020-646EA 577.02 USD
220020-646
Anti-FYB Mouse Monoclonal Antibody [clone: 460107]
Antibodies
Type: Primary
Antigen Symbol: FYB
Antigen Name: FYN binding protein
Antigen Synonyms: ADAP, PRO0823, SLAP-130, SLAP130
Clonality: Monoclonal
Clone: 460107
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR