Anti-LTBP2 Mouse Monoclonal Antibody [clone: 327318]

Supplier: R & D Systems
MAB3850
220020-644EA 577.02 USD
220020-644
Anti-LTBP2 Mouse Monoclonal Antibody [clone: 327318]
Antibodies
Type: Primary
Antigen: LTBP2
Clonality: Monoclonal
Clone: 327318
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR