Anti-NOTCH4 Rat Monoclonal Antibody [clone: 411913]

Supplier: R & D Systems
MAB3847
220020-641EA 577.02 USD
220020-641
Anti-NOTCH4 Rat Monoclonal Antibody [clone: 411913]
Antibodies
Type: Primary
Antigen: NOTCH4
Clonality: Monoclonal
Clone: 411913
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR