Anti-GRB2 Mouse Monoclonal Antibody [clone: 481021]

Supplier: R & D Systems
MAB3846
220020-640EA 577.02 USD
220020-640
Anti-GRB2 Mouse Monoclonal Antibody [clone: 481021]
Antibodies
Type: Primary
Antigen Symbol: GRB2
Antigen Name: growth factor receptor bound protein 2
Antigen Synonyms: -
Clonality: Monoclonal
Clone: 481021
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR