Anti-CFHR5 Mouse Monoclonal Antibody [clone: 390513]

Supplier: R & D Systems
MAB3845
220020-639EA 594.37 USD
220020-639
Anti-CFHR5 Mouse Monoclonal Antibody [clone: 390513]
Antibodies
Type: Primary
Antigen: CFHR5
Clonality: Monoclonal
Clone: 390513
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR