Anti-MAGED1 Mouse Monoclonal Antibody [clone: 353124]

Supplier: R & D Systems
MAB3835
220020-638EA 577.02 USD
220020-638
Anti-MAGED1 Mouse Monoclonal Antibody [clone: 353124]
Antibodies
Type: Primary
Antigen: MAGED1
Clonality: Monoclonal
Clone: 353124
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR