Anti-ERAP2 Mouse Monoclonal Antibody [clone: 3F5]

Supplier: R & D Systems
MAB3830
220020-633EA 600.15 USD
220020-633
Anti-ERAP2 Mouse Monoclonal Antibody [clone: 3F5]
Antibodies
Type: Primary
Antigen: ERAP2
Clonality: Monoclonal
Clone: 3F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR