Anti-ANGPTL3 Rat Monoclonal Antibody [clone: 38862]

Supplier: R & D Systems
MAB38291
220020-632EA 779.48 USD
220020-632
Anti-ANGPTL3 Rat Monoclonal Antibody [clone: 38862]
Antibodies
Type: Primary
Antigen: ANGPTL3
Clonality: Monoclonal
Clone: 38862
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR