Anti-ANGPTL3 Mouse Monoclonal Antibody [clone: 3727]

Supplier: R & D Systems
MAB3829
220020-631EA 577.02 USD
220020-631
Anti-ANGPTL3 Mouse Monoclonal Antibody [clone: 3727]
Antibodies
Type: Primary
Antigen: ANGPTL3
Clonality: Monoclonal
Clone: 3727
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR