Anti-STAB1 Mouse Monoclonal Antibody [clone: 840449]

Supplier: R & D Systems
MAB3825
102885-640EA 594.37 USD
102885-640
Anti-STAB1 Mouse Monoclonal Antibody [clone: 840449]
Antibodies
Type: Primary
Antigen: STAB1
Clonality: Monoclonal
Clone: 840449
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR