Anti-RELN Rat Monoclonal Antibody [clone: 397919]

Supplier: R & D Systems
MAB3820
220020-629EA 561.11 USD
220020-629
Anti-RELN Rat Monoclonal Antibody [clone: 397919]
Antibodies
Type: Primary
Antigen: RELN
Clonality: Monoclonal
Clone: 397919
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR