Anti-MCSF1R Rat Monoclonal Antibody [clone: 460630]

Supplier: R & D Systems
MAB38181
220020-627EA 594.37 USD
220020-627
Anti-MCSF1R Rat Monoclonal Antibody [clone: 460630]
Antibodies
Type: Primary
Antigen: MCSF1R
Clonality: Monoclonal
Clone: 460630
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR