Anti-ANPEP Mouse Monoclonal Antibody [clone: 4981]

Supplier: R & D Systems
MAB3815
220020-625EA 633.42 USD
220020-625
Anti-ANPEP Mouse Monoclonal Antibody [clone: 4981]
Antibodies
Type: Primary
Antigen: ANPEP
Clonality: Monoclonal
Clone: 4981
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR