Anti-CSF3R Mouse Monoclonal Antibody [clone: 38643]

Supplier: R & D Systems
MAB3811
220020-624EA 763.57 USD
220020-624
Anti-CSF3R Mouse Monoclonal Antibody [clone: 38643]
Antibodies
Type: Primary
Antigen: CSF3R
Clonality: Monoclonal
Clone: 38643
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR