Anti-MAFB Mouse Monoclonal Antibody [clone: 372239]

Supplier: R & D Systems
MAB3810
220020-623EA 600.15 USD
220020-623
Anti-MAFB Mouse Monoclonal Antibody [clone: 372239]
Antibodies
Simple Western

Type: Primary
Antigen: MAFB
Clonality: Monoclonal
Clone: 372239
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR