Anti-CSF3R Mouse Monoclonal Antibody [clone: 38660]

Supplier: R & D Systems
MAB381
220020-622EA 763.57 USD
220020-622
Anti-CSF3R Mouse Monoclonal Antibody [clone: 38660]
Antibodies
Type: Primary
Antigen: CSF3R
Clonality: Monoclonal
Clone: 38660
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR