Anti-MAFK Mouse Monoclonal Antibody [clone: 381923]

Supplier: R & D Systems
MAB3809
220020-621EA 577.02 USD
220020-621
Anti-MAFK Mouse Monoclonal Antibody [clone: 381923]
Antibodies
Type: Primary
Antigen: MAFK
Clonality: Monoclonal
Clone: 381923
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR