Anti-PRKAB2 Rat Monoclonal Antibody [clone: 376938]

Supplier: R & D Systems
MAB3808
220020-620EA 577.02 USD
220020-620
Anti-PRKAB2 Rat Monoclonal Antibody [clone: 376938]
Antibodies
Type: Primary
Antigen: PRKAB2
Clonality: Monoclonal
Clone: 376938
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR