Anti-EIF4B Rat Monoclonal Antibody [clone: 866049]

Supplier: R & D Systems
MAB3800
102885-638EA 561.11 USD
102885-638
Anti-EIF4B Rat Monoclonal Antibody [clone: 866049]
Antibodies
Type: Primary
Antigen: EIF4B
Clonality: Monoclonal
Clone: 866049
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR