Anti-TPSB2 Mouse Monoclonal Antibody [clone: 349414]

Supplier: R & D Systems
MAB37961
220020-618EA 600.15 USD
220020-618
Anti-TPSB2 Mouse Monoclonal Antibody [clone: 349414]
Antibodies
Type: Primary
Antigen: TPSB2
Clonality: Monoclonal
Clone: 349414
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR