Anti-CD4 Mouse Monoclonal Antibody [clone: 34915]

Supplier: R & D Systems
MAB3791
220020-616EA 763.57 USD
220020-616
Anti-CD4 Mouse Monoclonal Antibody [clone: 34915]
Antibodies
Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: 34915
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR