Anti-VAV1 Mouse Monoclonal Antibody [clone: 751411]

Supplier: R & D Systems
MAB37861
102885-636EA 594.37 USD
102885-636
Anti-VAV1 Mouse Monoclonal Antibody [clone: 751411]
Antibodies
Simple Western

Type: Primary
Antigen: VAV1
Clonality: Monoclonal
Clone: 751411
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR