Anti-VAV1 Mouse Monoclonal Antibody [clone: 591529]

Supplier: R & D Systems
MAB3786
220020-614EA 594.37 USD
220020-614
Anti-VAV1 Mouse Monoclonal Antibody [clone: 591529]
Antibodies
Type: Primary
Antigen: VAV1
Clonality: Monoclonal
Clone: 591529
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR