Anti-YY1 Mouse Monoclonal Antibody [clone: 854847]

Supplier: R & D Systems
MAB3784
102885-634EA 577.02 USD
102885-634
Anti-YY1 Mouse Monoclonal Antibody [clone: 854847]
Antibodies
Type: Primary
Antigen: YY1
Clonality: Monoclonal
Clone: 854847
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR