Anti-GC Mouse Monoclonal Antibody [clone: 359501]

Supplier: R & D Systems
MAB3778
220020-613EA 577.02 USD
220020-613
Anti-GC Mouse Monoclonal Antibody [clone: 359501]
Antibodies
Type: Primary
Antigen Symbol: GC
Antigen Name: group-specific component (vitamin D binding protein)
Antigen Synonyms: DBP, DBP/GC, GRD3, HEL-S-51, VDBG, VDBP
Clonality: Monoclonal
Clone: 359501
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR