Anti-FKBP1A Rat Monoclonal Antibody [clone: 422513]

Supplier: R & D Systems
MAB3777
220020-612EA 594.37 USD
220020-612
Anti-FKBP1A Rat Monoclonal Antibody [clone: 422513]
Antibodies
Type: Primary
Antigen Symbol: FKBP1A
Antigen Name: FK506 binding protein 1A, 12kDa
Antigen Synonyms: FKBP-12, FKBP-1A, FKBP1, FKBP12, PKC12, PKCI2, PPIASE
Clonality: Monoclonal
Clone: 422513
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR