Anti-HMOX1 Rat Monoclonal Antibody [clone: 412811]

Supplier: R & D Systems
MAB3776
220020-611EA 594.37 USD
220020-611
Anti-HMOX1 Rat Monoclonal Antibody [clone: 412811]
Antibodies
Type: Primary
Antigen: HMOX1
Clonality: Monoclonal
Clone: 412811
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR